Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 123170010
Price (USD)/ Each
90.65
Online exclusive
107.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Online exclusive
107.00 
Save 16.35 (15%)
Add to cart
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Catalog number123170010
Price (USD)/ Each
90.65
Online exclusive
107.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
Molecular Weight (g/mol)80.91
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Titration with NaOH>=47 %
Appearance (Color)Clear light yellow to orange
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only