Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number 123170025
Price (USD)/ Each
214.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
252.00 
Save 37.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
214.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
252.00 
Save 37.35 (15%)
Add to cart
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Catalog number123170025
Price (USD)/ Each
214.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
252.00 
Save 37.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
Molecular Weight (g/mol)80.91
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Appearance (Color)Clear light yellow to orange
Titration with NaOH>=47 %
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only