DynaMag™-2 Magnet
DynaMag™-2 Magnet
Invitrogen™
DynaMag™-2 Magnet
The DynaMag™-2 magnet is optimized for efficient magnetic separation of all types of Dynabeads™ (range 1–4.5 μm in diameter) inRead more
Have Questions?
Catalog NumberQuantity
12321D1 Each
Catalog number 12321D
Price (USD)
878.00
Each
-
Add to cart
Quantity:
1 Each
Price (USD)
878.00
Each
Add to cart
Ask our AI about this Product
The DynaMag™-2 magnet is optimized for efficient magnetic separation of all types of Dynabeads™ (range 1–4.5 μm in diameter) in small (less than 2 mL) sample volumes. The DynaMag™-2 magnet combines a strong magnetic attraction for efficient separations in standard microcentrifuge tubes with a flexible and smart ergonomic design.
• Optimal working volume: 10–2,000 μL
• Holds up to 16 standard 1.5–2 mL microcentrifuge tubes in numbered spaces
• Good control and visibility of your sample
• Top rack can be quickly removed from the magnet in the base, ready for resuspension, vortexing, rotation, or manual sample shaking
Flexible and ergonomic design
The magnet can be used with all types of Dynabeads™ for efficient isolation of proteins and nucleic acids. The sample rack can be easily removed from the base magnet for resuspension, vortexing, rotation, or manual sample shaking. A center pin in the rack ensures equal vortexing of all tubes. The rack can also be removed and used to store tubes. The 16 numbered sample spaces make this magnet ideal for parallel handling of multiple samples. With 8 tube positions on each side of the rack you can easily scale your protocols to accommodate 96-well plate reactions.
Product notes
• This magnet is not compatible with the MX4 Mixer
• The removable sample rack is also available as a separate item (Cat. No. 123-22D). Purchasing an additional sample rack minimizes the need to move tubes between racks. The rack can be placed on the bench, in the refrigerator, or in the incubator.
Learn more about Dynabeads™ products
• Find Dynabeads™ products for a whole range of applications.
• Find other magnets for Dynabeads™ separations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionMagnet
Quantity1 Each
Shipping ConditionRoom Temperature
Product LineDYNAL™, DynaMag™, Dynabeads™
Unit SizeEach
Contents & Storage
Contains one DynaMag™-2 magnet. Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link