Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 123240010
Price (USD)/ Each
58.65
Online exclusive
65.20 
Save 6.55 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
65.20 
Save 6.55 (10%)
Add to cart
Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number123240010
Price (USD)/ Each
58.65
Online exclusive
65.20 
Save 6.55 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
Molecular Weight (g/mol)136.08
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Chloride (Cl)=<200 ppm
Iron (Fe)=<10 ppm
Infrared spectrumConforms
Insoluble matter=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only