Thermo Scientific Chemicals

1,2-Pentanediol, 96%, Thermo Scientific Chemicals

Catalog number: 123292500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Pentanediol, 96%, Thermo Scientific Chemicals

Catalog number: 123292500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 123292500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5343-92-0
IUPAC Name
pentane-1,2-diol
Molecular Formula
C5H12O2
InChI Key
WCVRQHFDJLLWFE-UHFFFAOYNA-N
SMILES
CCCC(O)CO
Appearance (Form)
Liquid
GC
>=95.0 %
Infrared spectrum
Conforms
Refractive index
1.4380 to 1.4400 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow to orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text