Thermo Scientific Chemicals

1,2-Bis(2-aminophenoxy)-ethane-N,N,N'N'-tetraacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 123310050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

1,2-Bis(2-aminophenoxy)-ethane-N,N,N'N'-tetraacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 123310050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 123310050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
85233-19-8
IUPAC Name
tetrapotassium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Molecular Formula
C22H20K4N2O10
InChI Key
MAWIDOKQOLSOFT-UHFFFAOYSA-J
SMILES
[K+].[K+].[K+].[K+].[O-]C(=O)CN(CC([O-])=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC([O-])=O)CC([O-])=O
Infrared spectrum
Conforms
Appearance (Form)
Powder
Appearance (Color)
White to off-white
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text