Chemical Identifiers
CAS85233-19-8
IUPAC Nametetrapotassium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Molecular FormulaC22H20K4N2O10
InChI KeyMAWIDOKQOLSOFT-UHFFFAOYSA-J
SMILES[K+].[K+].[K+].[K+].[O-]C(=O)CN(CC([O-])=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC([O-])=O)CC([O-])=O
View more