DynaMag™-96 Bottom Magnet
DynaMag™-96 Bottom Magnet
Invitrogen™

DynaMag™-96 Bottom Magnet

96-well Magnet optimized for use with Dynabeads™ This magnet pulls beads to the bottom of each well. Compatible with PCR-stripsRead more
Have Questions?
Catalog NumberQuantity
12332D1 each
Catalog number 12332D
Price (USD)
1,192.65
Online Exclusive
1,236.00
Save 43.35 (4%)
Each
Add to cart
Quantity:
1 each
Price (USD)
1,192.65
Online Exclusive
1,236.00
Save 43.35 (4%)
Each
Add to cart
96-well Magnet optimized for use with Dynabeads™ This magnet pulls beads to the bottom of each well. Compatible with PCR-strips and 96-well plates (200ul) non-skirted and half-skirted. This magnet replaces the Dynal MPC™-96B.

When used in combination with the DynaMag™-96 Side (Product # 12331D) on an automation platform, the two magnets allow for significant volume changes through a workflow.
High wash volumes - DynaMag™-96 Side
Low Elution Volumes - DynaMag™-96 Bottom

Concentrates Beads at the bottom of each well.

Features 96 round magnets (one centered at the bottom of each well: to ensure identical sample handling at each position).

Allows for concentrating the sample by eluting in tiny volumes (∼10 μl).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionMagnet
For Use With (Equipment)DynaMag™
Quantity1 each
Shipping ConditionRoom Temperature
Product LineDynaMag™
Unit SizeEach
Contents & Storage
Contains one magnet. Store at room temperature.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection