Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals
Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals

Ammonium chloride, 99+%, H4ClN, CAS Number-12125-02-9, amchlor, ammoneric, ammoniumchlorid, ammonium chloride, ammonium chloride nh4 cl, ammoniumklorid, ammonium muriate, darammon, sal ammoniac, salmiac, 1kg, 99% min. (Argentometry), CHEBI:31206, White, 1.5200g/mL, 235-186-4, 15,13, 53.49, Pure
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number 123340250
Price (USD)/ Each
17.65
Online exclusive
20.70 
Save 3.05 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
17.65
Online exclusive
20.70 
Save 3.05 (15%)
Add to cart
Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals
Catalog number123340250
Price (USD)/ Each
17.65
Online exclusive
20.70 
Save 3.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameamine hydrochloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILESN.Cl
Molecular Weight (g/mol)53.49
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<10 ppm
Appearance (Color)White
Water=<0.1 % (K.F.)
Titration Argentometric>=99.0 %
Insoluble matter=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only