Thermo Scientific Chemicals

Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 123340251
25 kg, Fibre drum, Each
Thermo Scientific Chemicals

Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 123340251
25 kg, Fibre drum, Each
Quantity
Packaging
Catalog number: 123340251
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12125-02-9
IUPAC Name
amine hydrochloride
Molecular Formula
ClH4N
InChI Key
NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES
N.Cl
Iron (Fe)
=<10 ppm
Appearance (Color)
White
Water
=<0.1 % (K.F.)
Titration Argentometric
>=99.0 %
Insoluble matter
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text