Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Thermo Scientific Chemicals

Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2

CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 123350100
Price (USD)
475.00
Each
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
475.00
Each
Add to cart
Chemical Identifiers
CAS10035-04-8
IUPAC Namecalcium dihydrate dichloride
Molecular FormulaCaCl2H4O2
InChI KeyLLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Appearance (Form)Flakes
Titration Complexometric77 to 80 % (CaCl2)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection