Sodium Bicarbonate, 99+%, Extra Pure, Thermo Scientific Chemicals
Sodium Bicarbonate, 99+%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium Bicarbonate, 99+%, Extra Pure, Thermo Scientific Chemicals

Sodium bicarbonate, 99+%, CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 5kg, 20ppm max., 100ppm max., CHEBI:32139, White, 205-633-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number 123360050
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
Add to cart
Sodium Bicarbonate, 99+%, Extra Pure, Thermo Scientific Chemicals
Catalog number123360050
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HCl>=99.0 %
Chloride (Cl)=<100 ppm
Calcium (Ca)=<100 ppm
Potassium (K)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only