Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 123370250
25 kg, Fibre drum, Each
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 123370250
25 kg, Fibre drum, Each
Quantity
Packaging
Catalog number: 123370250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10102-17-7
IUPAC Name
disodium pentahydrate sulfanidesulfonate
Molecular Formula
H10Na2O8S2
InChI Key
PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
Appearance (Form)
Crystals
pH
6 to 8.4 (10 % at 20°C)
Calcium (Ca)
=<50 ppm
Chloride (Cl)
=<500 ppm
Sulfur (S)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text