Thermo Scientific Chemicals

Dicalite™ speed plus, Diatomaceous Filter-aid, Thermo Scientific Chemicals

Catalog number: 123380010
1 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Dicalite™ speed plus, Diatomaceous Filter-aid, Thermo Scientific Chemicals

Catalog number: 123380010
1 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 123380010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68855-54-9
Additional info
pH of a 10% slurry: 9 to 10.5
Additional info
retained on 40 mesh sieve: < 0.90 % w/w
Additional info
retained on 140 mesh sieve: < 10.0 % w/w
Appearance (Color)
White to off-white
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text