1x1 image pixel for data collection
AccuPrime™ Pfx DNA Polymerase
Upgrade to Invitrogen™ Platinum™ SuperFi™ II DNA Polymerase: >300X higher fidelity than Taq, versatile and now enables universal primer annealing.
AccuPrime&trade; <i>Pfx</i> DNA Polymerase
Invitrogen™

AccuPrime™ Pfx DNA Polymerase

AccuPrime Pfx DNA Polymerase is ideal for high-fidelity amplification of DNA fragments for downstream applications such as cloning and mutagenesis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
12344024200 Reactions
123440321000 Reactions
Catalog number 12344024
Price (USD)
406.90
Special offer
Online exclusive
Ends: 17-Jul-2025
626.00
Save 219.10 (35%)
Each
In stock
Add to cart
No. of Reactions:
200 Reactions
Request bulk or custom format
Price (USD)
406.90
Special offer
Online exclusive
Ends: 17-Jul-2025
626.00
Save 219.10 (35%)
Each
Add to cart
AccuPrime Pfx DNA Polymerase is ideal for high-fidelity amplification of DNA fragments for downstream applications such as cloning and mutagenesis. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcus species KOD with proofreading (3'→5' exonuclease) activity. Platinum anti-Pfx DNA polymerase antibodies inhibit polymerase activity, providing hot-start capabilities for improved PCR specificity (see figure). Thermostable AccuPrime accessory proteins enhance specific primer-template hybridization during every cycle of PCR, increasing specificity, yield, and robustness over Platinum Pfx alone.

Benefits of AccuPrime Pfx DNA Polymerase

  • Greater than 26 times higher fidelity than Taq DNA polymerase
  • Amplification of fragments up to 12 kb
  • Minimized PCR optimization

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • RT-PCR
  • High-fidelity PCR
  • Hot-start PCR

Notes

  • AccuPrime Pfx DNA Polymerase is especially effective where very high levels of fidelity are required.
  • The 10X AccuPrime Pfx reaction mix contains MgSO4, dNTPs, and thermostable proteins that enhance specific primer-template hybridization during each PCR cycle.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)26X
Hot StartBuilt-In Hot Start
No. of Reactions200 Reactions
OverhangBlunt
PolymeraseAccuPrime Pfx DNA Polymerase
Quantity200 Reactions
Reaction FormatSeparate Components
Shipping ConditionWet or Dry Ice
Size (Final Product)12 kb or less
Volume100 μL
Detection MethodPrimer-probe
For Use With (Application)Hot-start PCR, High-fidelity PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• AccuPrime Pfx DNA Polymerase (1 x 100 μL at 2.5 U/μL)
• 10X AccuPrime Pfx Reaction Mix (1 x 1 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -20°C.

Frequently asked questions (FAQs)

My oligonucleotide does not appear to be the right length when I checked by gel electrophoresis. Why is this?

Oligos should be run on a polyacrylamide gel containing 7 M urea and loaded with a 50% formamide solution to avoid compressions and secondary structures. Oligos of the same length and different compositions can electrophorese differently. dC's migrate fastest, followed by dA's, dT's, and then dG's. Oligos containing N's tend to run as a blurry band and generally have a problem with secondary structure.

The primers I am using worked for PCR initially, but over time, have stopped working. What happened?

Primers should be aliquoted for single use before PCR set-up. Heat just the aliquoted primers to 94 degrees for 1 min. Quick chill the primer on ice before adding to the PCR reaction. Some primers may anneal to themselves or curl up on themselves.

I don't see a pellet in my oligo tube order. Should I ask for a replacement?

The drying method dries the primer in a thin layer along the sidewalls of the tube instead of the bottom, therefore a pellet is not always visible and should still be ready to use.

There is a ball-shaped pellet at the bottom of my oligo tube. What is this and can I still use my oligo?

If the oligo was overheated, it will appear as a “ball”-shaped pellet attached to the bottom of the tube. This should not affect the quality of the oligo, and the oligo should be readily soluble in water.

There is a green color in my lyophilized oligo. Can I still use it?

If an oligo appears green in color, this is most likely due to ink falling into the tube. The oligo should still be fully functional. The color can be removed by doing an ethanol precipitation.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection