Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Thermo Scientific Chemicals

Catalog number: 123530025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Thermo Scientific Chemicals

Catalog number: 123530025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 123530025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2052-49-5, 108-88-3, 67-56-1
Titration with HClO4
0.09 to 0.11 M
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text