Sodium carbonate, 99.5%, extra pure, anhydrous, Thermo Scientific Chemicals
Sodium carbonate, 99.5%, extra pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium carbonate, 99.5%, extra pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 123670010
Price (USD)/ Each
84.65
Online exclusive
93.90 
Save 9.25 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
93.90 
Save 9.25 (10%)
Add to cart
Sodium carbonate, 99.5%, extra pure, anhydrous, Thermo Scientific Chemicals
Catalog number123670010
Price (USD)/ Each
84.65
Online exclusive
93.90 
Save 9.25 (10%)
-
Add to cart
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Granular powder
Titration with HCl99.5 to 100.5 % (On dried substance)
Loss on drying=<1 % (300°C, 2 hrs)
Heavy metals (as Pb)=<50 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Adsorbent
RUO – Research Use Only