Thermo Scientific Chemicals

Sodium carbonate, 99.5%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 123670025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Sodium carbonate, 99.5%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 123670025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 123670025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
497-19-8
IUPAC Name
disodium carbonate
Molecular Formula
CNa2O3
InChI Key
CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C([O-])=O
Appearance (Color)
White
Appearance (Form)
Granular powder
Titration with HCl
99.5 to 100.5 % (On dried substance)
Loss on drying
=<1 % (300°C, 2 hrs)
Heavy metals (as Pb)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Adsorbent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text