Thermo Scientific Chemicals

Diethyl (2-oxopropyl)phosphonate, 96%, Thermo Scientific Chemicals

Catalog number: 123690050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl (2-oxopropyl)phosphonate, 96%, Thermo Scientific Chemicals

Catalog number: 123690050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 123690050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1067-71-6
IUPAC Name
diethyl (2-oxopropyl)phosphonate
Molecular Formula
C7H15O4P
InChI Key
RSAFKRSMGOSHRK-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC(C)=O)OCC
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4325 to 1.4355 (20°C, 589 nm)
GC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text