Thermo Scientific Chemicals

8-Aminoquinoline, 98+%, Thermo Scientific Chemicals

Catalog number: 123710500
50 g, Each
Thermo Scientific Chemicals

8-Aminoquinoline, 98+%, Thermo Scientific Chemicals

Catalog number: 123710500
50 g, Each
Quantity
Catalog number: 123710500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
578-66-5
IUPAC Name
quinolin-8-amine
Molecular Formula
C9H8N2
InChI Key
WREVVZMUNPAPOV-UHFFFAOYSA-N
SMILES
NC1=C2N=CC=CC2=CC=C1
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
62°C to 65°C
Titration with HClO4
>=98.0 %
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text