Thermo Scientific Chemicals

Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals

Catalog number: 123770100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals

Catalog number: 123770100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 123770100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
311-89-7
IUPAC Name
tris(1,1,2,2,3,3,4,4,4-nonafluorobutyl)amine
Molecular Formula
C12F27N
InChI Key
RVZRBWKZFJCCIB-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Infrared spectrum
Conforms
Viscosity
2.3 to 3.3 cSt (at 25°C)
Additional info
residue: < 10 ug/ml
Additional info
dielectric strength: > 35 kV
Density
(25°C) 1.865 to 1.895 g/ml

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text