Chemical Identifiers
CAS311-89-7
IUPAC Nametris(1,1,2,2,3,3,4,4,4-nonafluorobutyl)amine
Molecular FormulaC12F27N
InChI KeyRVZRBWKZFJCCIB-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
View more