Chemical Identifiers
CAS6153-64-6
IUPAC Name(2Z,4S,4aR,5S,5aR,6S,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,6,10,11,12a-pentahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione dihydrate
Molecular FormulaC22H28N2O11
InChI KeyQRTMTZAUGTXZOA-ZGYDFYMOSA-N
SMILESO.O.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)C4=C(O)C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
View more