4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%, Thermo Scientific Chemicals
4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 123880050
Price (USD)/ Each
36.20
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
36.20
Add to cart
4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog number123880050
Price (USD)/ Each
36.20
-
Add to cart
Chemical Identifiers
CAS4940-39-0
IUPAC Name4-oxo-4H-chromene-2-carboxylate
Molecular FormulaC10H5O4
InChI KeyRVMGXWBCQGAWBR-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC(=O)C2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)White to beige to grey-brown
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only