Thermo Scientific Chemicals

Tetra-n-butylammonium tribromide, 98+%, Thermo Scientific Chemicals

Catalog number: 123971000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium tribromide, 98+%, Thermo Scientific Chemicals

Catalog number: 123971000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 123971000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
38932-80-8
IUPAC Name
N,N,N-trimethylanilinium
Molecular Formula
C9H14N
InChI Key
ZNEOHLHCKGUAEB-UHFFFAOYSA-N
SMILES
C[N+](C)(C)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
Melting point
71°C to 75°C
Appearance (Color)
Orange
Infrared spectrum
Conforms
Titration Iodometric
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text