4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, Thermo Scientific Chemicals
4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 g
Catalog number 123985000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-hydroxybenzene-1-sulfonate
Molecular FormulaC6H5O4S
InChI KeyFEPBITJSIHRMRT-UHFFFAOYSA-M
SMILESOC1=CC=C(C=C1)S([O-])(=O)=O
Molecular Weight (g/mol)173.16
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Appearance (Form)Powder or needles
Water13 to 17 % (K.F.)
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only