Thermo Scientific Chemicals

Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect., Thermo Scientific Chemicals

Catalog number: 124020025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect., Thermo Scientific Chemicals

Catalog number: 124020025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 124020025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
8042-47-5
Appearance (Form)
Oily liquid
Viscosity
13 to 19 mm²/s (40°C)
Appearance (Color)
Clear colorless
Refractive index
1.4620 to 1.4730 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For IR spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text