Thermo Scientific Chemicals

Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals

Catalog number: 124050050
5 L, Plastic Bottle, Each
Thermo Scientific Chemicals

Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals

Catalog number: 124050050
5 L, Plastic Bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 124050050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-09-2
IUPAC Name
dichloromethane
Molecular Formula
CH2Cl2
InChI Key
YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES
ClCCl
Appearance (Color)
Clear colorless
Residue after evaporation
=<0.05 %
Appearance (Form)
Liquid
Water
=<0.1 % (K.F.)
Stabilizer
Approx. 0.2 % Ethanol

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text