Thermo Scientific Chemicals

Silver nitrate, 0.1 N standard solution, Thermo Scientific Chemicals

Catalog number: 124250010
1 L, HDPE Bottle, Each
Thermo Scientific Chemicals

Silver nitrate, 0.1 N standard solution, Thermo Scientific Chemicals

Catalog number: 124250010
1 L, HDPE Bottle, Each
Quantity
Packaging
Catalog number: 124250010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7761-88-8, 7732-18-5
Appearance (Form)
Solution
Strength
0.0998 to 0.1002 N (20°C)
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text