Thermo Scientific Chemicals

Sodium thiosulfate, 0.1 N standard solution, Thermo Scientific Chemicals

Catalog number: 124270250
25 L, Plastic Drum, Each
Thermo Scientific Chemicals

Sodium thiosulfate, 0.1 N standard solution, Thermo Scientific Chemicals

Catalog number: 124270250
25 L, Plastic Drum, Each
Quantity
Packaging
Catalog number: 124270250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7772-98-7, 7732-18-5
Appearance (Form)
Solution
Appearance (Color)
Clear colorless
Titration Iodimetric
0.0950 to 0.1050 N (20°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent, Volumetric standard solution
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text