Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

IMDM

Catalog number: 12440046
Have Questions?
Gibco™

IMDM

Catalog number: 12440046
Have Questions?
Catalog Number
12440046
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1,000 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
124400461,000 mL
-
12440053500 mL
-
1244006110 x 500 mL
-
124400796 x 1,000 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:
WithWithout
• L-glutamine• α-thioglycerol
• Phenol Red• 2-mercaptoethanol


The complete formulation is available.

Using IMDM
IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Specifications

Cell Line
Jurkat, COS-7, and macrophage cells
Classification
Animal Origin-free
Concentration
1 X
Form
Liquid
Product Type
IMDM (Iscove's Modified Dulbecco's Medium)
Shelf Life
12 Months From Date of Manufacture
Sterility
Sterile-filtered
With Additives
High Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
Without Additives
No α-Thioglycerol, No 2-Mercaptoethanol
Green Features
Sustainable packaging
Manufacturing Quality
cGMP-compliant under the ISO 13485 standard
Product Line
Gibco™
Quantity
1000 mL
Shipping Condition
Room Temperature

Contents & Storage

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text