Nitric acid, 70%, Thermo Scientific Chemicals
Nitric acid, 70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitric acid, 70%, Thermo Scientific Chemicals

May darken on storage
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number 124650010
Price (USD)/ Each
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH67 to 70 %
Appearance (Form)Clear liquid
AppearanceFree from suspended matter or sediment
Color scale=<30 APHA
AppearanceMay darken on storage
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only