Thermo Scientific Chemicals

Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals

Catalog number: 124660010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals

Catalog number: 124660010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 124660010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7697-37-2
IUPAC Name
nitric acid
Molecular Formula
HNO3
InChI Key
GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES
O[N+]([O-])=O
Chloride (Cl)
=<0.5 ppm
Copper (Cu)
=<0.05 ppm
Cadmium (Cd)
=<0.01 ppm
Arsenic (As)
=<0.01 ppm
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text