Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
25 L
Packaging:
View all
Glass Bottle
Plastic bottle
Catalog number 124660025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.5 ppm
Copper (Cu)=<0.05 ppm
Cadmium (Cd)=<0.01 ppm
Arsenic (As)=<0.01 ppm
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only