Methanol, 99.9%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals
Methanol, 99.9%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol, 99.9%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 124790010
Price (USD)/ Each
49.65
Online exclusive
58.70 
Save 9.05 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
58.70 
Save 9.05 (15%)
Add to cart
Methanol, 99.9%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals
Catalog number124790010
Price (USD)/ Each
49.65
Online exclusive
58.70 
Save 9.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
Molecular Weight (g/mol)32.04
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
UVat 240 nm A: =<0.10
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.85 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only