Magnesium sulfate heptahydrate, 99+%, extra pure
Magnesium sulfate heptahydrate, 99+%, extra pure
Magnesium sulfate heptahydrate, 99+%, extra pure
Magnesium sulfate heptahydrate, 99+%, extra pure
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, 99+%, extra pure

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 124900010
Price (USD)
73.65
Online Exclusive
81.70
Save 8.05 (10%)
Each
In stock
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
73.65
Online Exclusive
81.70
Save 8.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS10034-99-8
IUPAC Namemagnesium(2+) heptahydrate sulfate
Molecular FormulaH14MgO11S
InChI KeyWRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying48.0 % to 52.0 % (450°C)
Appearance (Form)Crystalline powder or crystals
Heavy metals (as Pb)=<10 ppm
Appearance (Color)White
Titration Complexometric>=99.0 % (on dry substance)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472097Certificate of AnalysisMay 17, 2025124900025, 124900100, 124900010
A0467197Certificate of AnalysisNov 11, 202412490001, 124900025, 12490017, 124900100, 124900010, 12490003, 12490025
A0463867Certificate of AnalysisJun 18, 202412490001, 124900025, 12490017, 124900100, 124900010, 12490003, 12490025
A0457808Certificate of AnalysisNov 14, 202312490001, 124900025, 12490017, 124900100, 124900010, 12490003, 12490025
A0429628Certificate of AnalysisNov 06, 2023124900025, 124900100, 124900010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection