Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 124910025
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
Add to cart
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Catalog number124910025
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5949-29-1
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular FormulaC6H10O8
InChI KeyYASYEJJMZJALEJ-UHFFFAOYSA-N
SMILESO.OC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH99.4 to 101.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
RUO – Research Use Only