Advanced DMEM
Save up to 29% on your order of cell culture products. View promotion details >
Advanced DMEM
Gibco™

Advanced DMEM

Advanced DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12491015Promo Image500 mL
12491023Promo Image10 x 500 mL
Catalog number 12491015
Price (USD)
50.65
Online Exclusive
55.50
Save 4.85 (9%)
Each
In stock
Add to cart
Quantity:
500 mL
Customize this product
Price (USD)
50.65
Online Exclusive
55.50
Save 4.85 (9%)
Each
Add to cart

Advanced DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic DMEM, serum supplementation can be reduced by 50–90% with no change in growth rate or morphology. Cells successfully cultured in Advanced DMEM, with no adaptation, include MDBK, HepG2, COS-7, A549, MDCK, WI-38, and Vero.

This Advanced DMEM is manufactured as follows:

With: High Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red

Without: L-glutamine

The complete formulation is available.

Using Advanced DMEM

Advanced DMEM is unique from other media due to the addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride. Advanced DMEM requires supplementation with 1–5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineMDBK, HepG2, COS-7, A549, MDCK, WI-38, and Vero
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco™
Product TypeDMEM (Dulbecco's Modified Eagle Medium)
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin
FormLiquid
Serum LevelReduced Serum
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesHigh Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
Without AdditivesNo Glutamine, No HEPES
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Media Formulations

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link