Search Thermo Fisher Scientific
Type | Sample Plug-in Cone |
Description | Plug-in Cone Kit, For Magnetic Holder (As Upgrade, Factory Installed Since 2009) |
For Use With (Equipment) | ELEMENT GD GD-MS Instrument |
Unit Size | Each |
Type | Sample Ni Cone |
Description | Ni Cone, For Fixing with 3 Screws (Not for Magnetic Holder) |
For Use With (Equipment) | ELEMENT GD GD-MS Instrument |
Unit Size | Each |
Type | Sample Plug-in Cone |
Description | Plug-in Cone Stainless Steel, For Magnetic Holder, Quantity: Singles |
For Use With (Equipment) | ELEMENT GD GD-MS Instrument |
Unit Size | Each |
Type | Sample Plug-in Cone |
Description | Plug-in Cone Graphite Uncoated, For Magnetic Holder, Quantity: Singles |
For Use With (Equipment) | ELEMENT GD GD-MS Instrument |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
1249180 | Each | Plug-in Cone Kit, For Magnetic Holder (As Upgrade, Factory Installed Since 2009) | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1186630 | Each | Ni Cone, For Fixing with 3 Screws (Not for Magnetic Holder) | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1239150 | Each | Plug-in Cone Stainless Steel, For Magnetic Holder, Quantity: Singles | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1239850 | Each | Plug-in Cone Graphite Uncoated, For Magnetic Holder, Quantity: Singles | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|