N-(2-Hydroxyethyl)phthalimide, 99%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)phthalimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Hydroxyethyl)phthalimide, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 124920250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3891-07-4
IUPAC Name2-(2-hydroxyethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC10H9NO3
InChI KeyMWFLUYFYHANMCM-UHFFFAOYSA-N
SMILESOCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
HPLC>=98.5 % (on dry substance)
Appearance (Form)Crystalline powder
Melting point126°C to 129°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only