Thermo Scientific Chemicals

1-(4-Nitrophenyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: 124940050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-(4-Nitrophenyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: 124940050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 124940050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6269-89-2
IUPAC Name
1-(4-nitrophenyl)piperazine
Molecular Formula
C10H13N3O2
InChI Key
VWOJSRICSKDKAW-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(C=C1)N1CCNCC1
Appearance (Color)
Yellow to ochre
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
130°C to 134°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text