1x1 image pixel for data collection
Sodium phosphate dibasic dihydrate, 98+%, for analysis
Sodium phosphate dibasic dihydrate, 98+%, for analysis
Sodium phosphate dibasic dihydrate, 98+%, for analysis
Sodium phosphate dibasic dihydrate, 98+%, for analysis
Thermo Scientific Chemicals

Sodium phosphate dibasic dihydrate, 98+%, for analysis

CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1249900505 kgPlastic drum
Catalog number 124990050
Price (USD)
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS10028-24-7
IUPAC Namedisodium dihydrate hydrogen phosphate
Molecular FormulaH5Na2O6P
InChI KeyKDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILESO.O.[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White
Titration excess HCl98.0 to 102.0 %
Insoluble matter=<0.3 % (in water)
pH8.4 to 9.6 (1% in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link