Sodium bisulfate monohydrate, 99%, for analysis
Sodium bisulfate monohydrate, 99%, for analysis
Sodium bisulfate monohydrate, 99%, for analysis
Sodium bisulfate monohydrate, 99%, for analysis
Thermo Scientific Chemicals

Sodium bisulfate monohydrate, 99%, for analysis

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 125005000
Price (USD)
136.65
Special offer
Online exclusive
Ends: 26-Sep-2025
168.00
Save 31.35 (19%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 26-Sep-2025
168.00
Save 31.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS10034-88-5
IUPAC Namesodium hydrate hydrogen sulfate
Molecular FormulaH3NaO5S
InChI KeyJXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILESO.[Na+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Adhering crystals
Titration with NaOH>=98.5 %
Chloride (Cl)=<0.0005 %
Iron (Fe)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link