Thermo Scientific Chemicals

Sodium sulfate decahydrate, extra pure, Thermo Scientific Chemicals

Catalog number: 125012500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium sulfate decahydrate, extra pure, Thermo Scientific Chemicals

Catalog number: 125012500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 125012500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7727-73-3
IUPAC Name
disodium decahydrate sulfate
Molecular Formula
H20Na2O14S
InChI Key
RSIJVJUOQBWMIM-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O
Appearance (Form)
Crystalline powder or crystals
Calcium (Ca)
=<0.01 %
Potassium (K)
=<0.01 %
Appearance (Color)
White
pH
5.2 to 9.2 (5% solution)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text