Thermo Scientific Chemicals

L-Leucine methyl ester hydrochloride, 98%

Catalog number: 125130250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

L-Leucine methyl ester hydrochloride, 98%

Catalog number: 125130250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 125130250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7517-19-3
IUPAC Name
methyl (2S)-2-amino-4-methylpentanoate hydrochloride
Molecular Formula
C7H16ClNO2
InChI Key
DODCBMODXGJOKD-HQFIOVGONA-N
SMILES
Cl.COC(=O)[C@@H](N)CC(C)C
Appearance (Form)
Powder or crystals
Titration with HClO4
>=97.5 %
Infrared spectrum
Conforms
Appearance (Color)
White to yellowish-white
Specific optical rotation
+18.5° to +21.5° (20°C, 589 nm) (c=4.5, methanol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text