Thermo Scientific Chemicals

Levulinic Acid, 98+%, Thermo Scientific Chemicals

Catalog number: 125140010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Levulinic Acid, 98+%, Thermo Scientific Chemicals

Catalog number: 125140010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 125140010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123-76-2
IUPAC Name
4-oxopentanoic acid
Molecular Formula
C5H8O3
InChI Key
JOOXCMJARBKPKM-UHFFFAOYSA-N
SMILES
CC(=O)CCC(O)=O
Appearance
May darken on storage
Titration with NaOH
>=98.0 %
Appearance (Color)
Colorless to white to yellow to orange
Refractive index
1.4390 to 1.4430 (20°C, 589 nm)
Appearance (Form)
Liquid after melting or solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text