Appearance (Color)Colorless to white to yellow to orange
Refractive index1.4390 to 1.4430 (20°C, 589 nm)
Appearance (Form)Liquid after melting or solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.