1x1 image pixel for data collection
2,6-Lutidine, 99%
2,6-Lutidine, 99%
2,6-Lutidine, 99%
Thermo Scientific Chemicals

2,6-Lutidine, 99%

CAS: 108-48-5 | C7H9N | 107.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 125192500
Price (USD)
63.65
Online Exclusive
70.00
Save 6.35 (9%)
Each
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.00
Save 6.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS108-48-5
IUPAC Name2,6-dimethylpyridine
Molecular FormulaC7H9N
InChI KeyOISVCGZHLKNMSJ-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4966 to 1.4986 (20°C, 589 nm)
AppearanceMay darken on storage
Water=<0.2 %
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link