Thermo Scientific Chemicals

L(+)-Lysine Monohydrochloride, ≥99%

Catalog number: 125220050
5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

L(+)-Lysine Monohydrochloride, ≥99%

Catalog number: 125220050
5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 125220050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
657-27-2
IUPAC Name
(2S)-2,6-diaminohexanoic acid hydrochloride
Molecular Formula
C6H15ClN2O2
InChI Key
BVHLGVCQOALMSV-JEDNCBNOSA-N
SMILES
Cl.NCCCC[C@H](N)C(O)=O
Sulfate (SO4)
=<300 ppm
Appearance (Form)
Crystalline powder or crystals
Loss on drying
=<0.4 % (105°C, 3 h)
Appearance (Color)
White
Titration with HClO4
98.5 to 101.0 % (on dried substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text