L(+)-Lysine Monohydrochloride, ≥99%
L(+)-Lysine Monohydrochloride, ≥99%
Thermo Scientific Chemicals

L(+)-Lysine Monohydrochloride, ≥99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 125222500
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.30 
Save 9.65 (14%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.30 
Save 9.65 (14%)
Add to cart
L(+)-Lysine Monohydrochloride, ≥99%
Catalog number125222500
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
68.30 
Save 9.65 (14%)
-
Add to cart
Chemical Identifiers
CAS657-27-2
IUPAC Name(2S)-2,6-diaminohexanoic acid hydrochloride
Molecular FormulaC6H15ClN2O2
InChI KeyBVHLGVCQOALMSV-JEDNCBNOSA-N
SMILESCl.NCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Loss on drying=<0.4 % (105°C, 3 h)
Appearance (Color)White
Titration with HClO498.5 to 101.0 % (on dried substance)
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only